Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100B Rabbit anti-Bovine, Human, Mouse, Rat, Clone-S100B/1706R, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP254426200UG
Description
IHC-P: Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer pH 6.0 for 10-20 min followed by cooling at RT for 20 minutesSpecifications
S100B | |
Monoclonal | |
1 mg/mL | |
Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.25 - 0.5 ug/ml, CyTOF-ready | |
beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta | |
Rabbit | |
11 kDa | |
200 μg | |
Apoptosis, Biologically Active Proteins, Cytoskeleton Markers, Neuroscience, Stem Cells | |
6285 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
S100B/1706R | |
Unconjugated | |
PBS with No Preservative | |
S100B | |
Recombinant full-length human S100B protein | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction