Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100B Rabbit anti-Bovine, Human, Mouse, Rat, Clone-S100B/1706R, Novus Biologicals™

Rabbit Monoclonal Antibody
$658.00
Specifications
Antigen | S100B |
---|---|
Clone | S100B/1706R |
Concentration | 1 mg/mL |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB254426100
![]() |
Novus Biologicals
NBP254426100UG |
100 μg |
Each for $658.00
|
|
|||||
NB254426200
![]() |
Novus Biologicals
NBP254426200UG |
200 μg | N/A | N/A | N/A | ||||
Description
S100B Monoclonal specifically detects S100B in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
S100B | |
1 mg/mL | |
Monoclonal | |
Rabbit | |
Apoptosis, Biologically Active Proteins, Cytoskeleton Markers, Neuroscience, Stem Cells | |
PBS with No Preservative | |
6285 | |
Recombinant full-length human S100B protein | |
Primary | |
11 kDa |
S100B/1706R | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Bovine | |
beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta | |
S100B | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title