Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100P Antibody (S100P/4386R), Biotin, Novus Biologicals™
SDP

Catalog No. NB390810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB390810 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB390810 Supplier Novus Biologicals Supplier No. NBP308769B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

S100P Monoclonal antibody specifically detects S100P in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen S100P
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S100P/4386R
Conjugate Biotin
Formulation PBS
Gene Alias MIG9, migration-inducing gene 9, Protein S100-E, S100 calcium binding protein P, S100 calcium-binding protein Pprotein S100-P, S100E
Host Species Rabbit
Immunogen Recombinant fragment corresponding to C-terminal of human S100P protein (Uniprot: P25815)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6286
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.