Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S1P1/EDG-1 Antibody (218713), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB366729 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB366729 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB366729 Supplier Novus Biologicals Supplier No. FAB2016J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

S1P1/EDG-1 Monoclonal specifically detects S1P1/EDG-1 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen S1P1/EDG-1
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 218713
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD363, CD363 antigen, CHEDG1, D1S3362, EDG-1, EDG1edg-1, Endothelial differentiation G-protein coupled receptor 1, endothelial differentiation, sphingolipid G-protein-coupled receptor, 1, FLJ58121, S1P receptor 1, S1P receptor Edg-1, s1p1, S1P1ECGF1, sphingosine 1-phosphate receptor 1, sphingosine 1-phosphate receptor EDG1, Sphingosine 1-phosphate receptor Edg-1, sphingosine-1-phosphate receptor 1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human S1P1/EDG-1, Met1-Ser382, Accession # P21453
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, GPCR, Lipid and Metabolism, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1901
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.