Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S1P1/EDG-1 Antibody (MM0045-21L9) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018047 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018047 0.025 mg
NB11093513 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018047 Supplier Novus Biologicals Supplier No. NB110935130.025MG

Mouse Monoclonal Antibody has been used in 2 publications

S1P1/EDG-1 Monoclonal specifically detects S1P1/EDG-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.

Specifications

Antigen S1P1/EDG-1
Applications CyTOF, Immunohistochemistry, Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0045-21L9
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:1000, Flow Cytometry 1:50-1:200, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50, CyTOF-ready
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. P21453
Gene Alias CD363, CD363 antigen, CHEDG1, D1S3362, EDG-1, EDG1edg-1, Endothelial differentiation G-protein coupled receptor 1, endothelial differentiation, sphingolipid G-protein-coupled receptor, 1, FLJ58121, S1P receptor 1, S1P receptor Edg-1, s1p1, S1P1ECGF1, sphingosine 1-phosphate receptor 1, sphingosine 1-phosphate receptor EDG1, Sphingosine 1-phosphate receptor Edg-1, sphingosine-1-phosphate receptor 1
Gene Symbols S1PR1
Host Species Mouse
Immunogen human recombinant EDG-1 protein
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 1901
Test Specificity This recognizes human EDG-1 in western blot.
Reconstitution Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.