Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S1P2/EDG-5/S1PR2 Antibody, Novus Biologicals™
SDP

Catalog No. NB158237 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB158237 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB158237 Supplier Novus Biologicals Supplier No. NBP314465
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

S1P2/EDG-5/S1PR2 Polyclonal antibody specifically detects S1P2/EDG-5/S1PR2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen S1P2/EDG-5/S1PR2
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias AGR16, EDG5, EDG-5, endothelial differentiation G-protein coupled receptor 5, endothelial differentiation, sphingolipid G-protein-coupled receptor, 5, Gpcr13, H218, LPB2, S1P receptor 2, S1P receptor Edg-5, S1P receptor EDG5, S1P2, sphingosine 1-phosphate receptor 2, sphingosine 1-phosphate receptor Edg-5, sphingosine-1-phosphate receptor 2
Host Species Rabbit
Immunogen Synthetic peptide - KLH conjugated
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 9294
Target Species Human
Content And Storage Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Product Type Antibody
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.