Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S1P3/EDG-3 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP224762

 View more versions of this product

Catalog No. NBP224762


Only null left
Add to Cart

Description

Description

S1P3/EDG-3 Polyclonal specifically detects S1P3/EDG-3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

S1P3/EDG-3
Polyclonal
Unconjugated
PBS with 0.05% Sodium Azide
EDG-3, EDG3FLJ37523, Endothelial differentiation G-protein coupled receptor 3, endothelial differentiation, sphingolipid G-protein-coupled receptor, 3, FLJ93220, G protein-coupled receptor, endothelial differentiation gene-3, LPB3, MGC71696, S1P receptor 3, S1P receptor Edg-3, S1P receptor EDG3, S1P3, sphingosine 1-phosphate receptor 3, Sphingosine 1-phosphate receptor Edg-3, sphingosine-1-phosphate receptor 3
Rabbit
Protein G purified
RUO
Primary
The amino acid sequence used as immunogen is 100% homologous in human and 95% homologous in mouse.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry (Paraffin)
1.0 mg/mL
Western Blot 1.0-2.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Q99500
S1PR3
A portion of amino acids 300-350 of human Sphingolipid Receptor Edg3/S1P3 was used as the immunogen.
0.1 mg
Cell Cycle and Replication, Cytoskeleton Markers, GPCR, Lipid and Metabolism, Signal Transduction
1903
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only