Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S1P3/EDG-3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$207.00 - $499.50
Specifications
Antigen | S1P3/EDG-3 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1.0-2.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224762SS
![]() |
Novus Biologicals
NBP224762SS |
0.025 mg |
Each for $207.00
|
|
|||||
NBP224762
![]() |
Novus Biologicals
NBP224762 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
S1P3/EDG-3 Polyclonal specifically detects S1P3/EDG-3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
S1P3/EDG-3 | |
Western Blot 1.0-2.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
Q99500 | |
1903 | |
A portion of amino acids 300-350 of human Sphingolipid Receptor Edg3/S1P3 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Cytoskeleton Markers, GPCR, Lipid and Metabolism, Signal Transduction | |
PBS with 0.05% Sodium Azide | |
EDG-3, EDG3FLJ37523, Endothelial differentiation G-protein coupled receptor 3, endothelial differentiation, sphingolipid G-protein-coupled receptor, 3, FLJ93220, G protein-coupled receptor, endothelial differentiation gene-3, LPB3, MGC71696, S1P receptor 3, S1P receptor Edg-3, S1P receptor EDG3, S1P3, sphingosine 1-phosphate receptor 3, Sphingosine 1-phosphate receptor Edg-3, sphingosine-1-phosphate receptor 3 | |
S1PR3 | |
IgG | |
Protein G purified | |
The amino acid sequence used as immunogen is 100% homologous in human and 95% homologous in mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title