Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S1P5/EDG-8 Antibody (1196A), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB345870 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345870 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345870 Supplier Novus Biologicals Supplier No. FAB9084MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

S1P5/EDG-8 Monoclonal specifically detects S1P5/EDG-8 in Mouse samples. It is validated for Flow Cytometry, Western Blot.

Specifications

Antigen S1P5/EDG-8
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 1196A
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Edg-8, EDG8S1P receptor Edg-8, Endothelial differentiation G-protein-coupled receptor 8, endothelial differentiation, sphingolipid G-protein-coupled receptor, 8, S1P receptor 5, S1P5, sphingosine 1-phosphate receptor 5, sphingosine 1-phosphate receptor EDG8, Sphingosine 1-phosphate receptor Edg-8, sphingosine-1-phosphate receptor 5, SPPR-1, SPPR-2
Host Species Rabbit
Immunogen Mouse S1P5/EDG-8 peptide corresponding to the N-terminal extracellular sequence, Accession # Q91X56
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 53637
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.