Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S1P5/EDG-8 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224712SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224712SS 0.025 mg
NBP224712 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224712SS Supplier Novus Biologicals Supplier No. NBP224712SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

S1P5/EDG-8 Polyclonal specifically detects S1P5/EDG-8 in Human, Mouse, Rat, Canine, Primate, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen S1P5/EDG-8
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml
Formulation 0.2 ml PBS with 0.05% Sodium Azide
Gene Accession No. Q9H228
Gene Alias Edg-8, EDG8S1P receptor Edg-8, Endothelial differentiation G-protein-coupled receptor 8, endothelial differentiation, sphingolipid G-protein-coupled receptor, 8, S1P receptor 5, S1P5, sphingosine 1-phosphate receptor 5, sphingosine 1-phosphate receptor EDG8, Sphingosine 1-phosphate receptor Edg-8, sphingosine-1-phosphate receptor 5, SPPR-1, SPPR-2
Gene Symbols S1PR5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 15-55 of human Edg8 was used as the immunogen, GenBank no NP_110387.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 53637
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a) and chimpanzee, 94% homologous in rhesus monkey, 84% homologous in cow and wild boar and 73% homologous in horse.
Target Species Human, Mouse, Rat, Canine, Primate, Xenopus species
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.