Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S1P5/EDG-8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$206.50 - $499.50
Specifications
Antigen | S1P5/EDG-8 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224712SS
![]() |
Novus Biologicals
NBP224712SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP224712
![]() |
Novus Biologicals
NBP224712 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
S1P5/EDG-8 Polyclonal specifically detects S1P5/EDG-8 in Human, Mouse, Rat, Canine, Primate, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
S1P5/EDG-8 | |
Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 5-10 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Canine, Primate, Xenopus species | |
Q9H228 | |
53637 | |
A synthetic peptide corresponding to amino acids 15-55 of human Edg8 was used as the immunogen, GenBank no NP_110387. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
GPCR | |
0.2 ml PBS with 0.05% Sodium Azide | |
Edg-8, EDG8S1P receptor Edg-8, Endothelial differentiation G-protein-coupled receptor 8, endothelial differentiation, sphingolipid G-protein-coupled receptor, 8, S1P receptor 5, S1P5, sphingosine 1-phosphate receptor 5, sphingosine 1-phosphate receptor EDG8, Sphingosine 1-phosphate receptor Edg-8, sphingosine-1-phosphate receptor 5, SPPR-1, SPPR-2 | |
S1PR5 | |
IgG | |
Protein G purified | |
The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a) and chimpanzee, 94% homologous in rhesus monkey, 84% homologous in cow and wild boar and 73% homologous in horse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title