Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ S4
GREENER_CHOICE

Catalog No. p-7110954
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

High affinity and selective CA IX and CA XII inhibitor

S4 is a high affinity and selective CA IX and CA XII inhibitor (Ki values are 2, 7, 547 and 5600 nM for CA XII, IX, II and I, respectively). Inhibits migration and proliferation of breast cancer cells in vitro. Inhibits lung metastasis in mice bearing breast tumor xenografts, without affecting primary tumor.

Specifications

Chemical Name or Material 4-[[[(3,5-Dimethylphenyl)amino]carbonyl]amino]phenyl sulfamate
CAS 1330061-67-0
Quantity 10 mg
Target Carbonic Anhydrase Inhibitors
Molecular Formula C15H17N3O4S
Formula Weight 335.38
Purity >99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.