Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S5a/Angiocidin Antibody (CPTC-PSMD4-3), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB387646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387646 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387646 Supplier Novus Biologicals Supplier No. NBP308271AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

S5a/Angiocidin Monoclonal antibody specifically detects S5a/Angiocidin in Human samples. It is validated for Western Blot, Protein Array

Specifications

Antigen S5a/Angiocidin
Applications Western Blot, Protein Array
Classification Monoclonal
Clone CPTC-PSMD4-3
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias 26S proteasome non-ATPase regulatory subunit 4;AF;AF-1;ASF;MCB1;PSMD4;pUB-R5;Rpn10;S5A
Host Species Mouse
Immunogen Recombinant full-length human S5a/Angiocidin protein (Uniprot: P55036)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5710
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.