Learn More
Description
This antibody detects the Cas9 of Staphylococcus aureus (SaCas9). It does not detect the SpCas9.
Specifications
Specifications
| Antigen | Sa Cas9 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.23 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene Alias | Cas9, Sa Cas9, SaCas9 |
| Host Species | Rabbit |
| Immunogen | Sa Cas9 Fusion Protein Ag25669 |
| Purification Method | Antigen Affinity Chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
