Learn More
Sa Cas9 Rabbit anti-Bacteria, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | Sa Cas9 |
|---|---|
| Concentration | 0.23 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-114-2786
![]()
|
Proteintech Group Inc
269271AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody detects the Cas9 of Staphylococcus aureus (SaCas9). It does not detect the SpCas9.
Cas9 (CRISPR associated protein 9) is an RNA-guided DNA endonuclease enzyme associated with the CRISPR (Clustered Regularly Interspaced Short Palindromic Repeats) adaptive immunity system in Streptococcus pyogenes (Sp).Specifications
| Sa Cas9 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Bacteria | |
| Cas9, Sa Cas9, SaCas9 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
| 0.23 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Sa Cas9 Fusion Protein Ag25669 | |
| Primary | |
| -20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.