Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SA1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB292208 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292208 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292208 Supplier Novus Biologicals Supplier No. NB100299PE
Add to Cart
Add to Cart

Goat Polyclonal Antibody

SA1 Polyclonal specifically detects SA1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot.

Specifications

Antigen SA1
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation PBS
Gene Alias cohesin subunit SA-1, nuclear protein stromal antigen 1, SA1, stromal antigen 1
Host Species Goat
Immunogen A 35 amino acid synthetic peptide, which represented a portion of human Stromal Antigen 1 encoded in part by exons 35 and 36 (LocusLink ID 10274).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, DNA Repair, Mitotic Regulators
Primary or Secondary Primary
Gene ID (Entrez) 10274
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.