Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SA1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB292212 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292212 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292212 Supplier Novus Biologicals Supplier No. NB100298PCP
Add to Cart
Add to Cart

Goat Polyclonal Antibody

SA1 Polyclonal antibody specifically detects SA1 in Human, Mouse, Invertebrate samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen SA1
Applications Western Blot, Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation PBS
Gene Alias cohesin subunit SA-1, nuclear protein stromal antigen 1, SA1, stromal antigen 1
Host Species Goat
Immunogen A 36 amino acid synthetic peptide, which represented a portion of human Stromal Antigen 1 encoded in part by exons 5 and 6 (LocusLink ID 10274).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, DNA Repair, Mitotic Regulators
Primary or Secondary Primary
Gene ID (Entrez) 10274
Target Species Human, Mouse, Invertebrate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.