Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saccharin Sodium (Powder/USP/FCC), Fisher Chemical™

Catalog No. s394212 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S394-212 2.5 kg Poly Bottle
S394-500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S394-212
Only null left
Add to Cart

Chemical Identifiers

CAS 128-44-9
Molecular Formula C7H9NNaO5S
Molecular Weight (g/mol) 242.201
MDL Number MFCD00013092
InChI Key UQKRDMFNUSMUCB-UHFFFAOYSA-N
Synonym saccharin sodium dihydrate, saccharin sodium, sodium saccharin, saccharin soluble, saccharin sodium salt hydrate, 1,2-benzisothiazol-3 2h-one, 1,1-dioxide, sodium salt, dihydrate, o-sulfobenzimide sodium salt, 2-sulfobenzoic acid imide sodium salt, saccharin sodium salt dihydrate, c7h4nnao3s.2h2o
PubChem CID 46942257
IUPAC Name 1,1-dioxo-1,2-benzothiazol-3-one;sodium;dihydrate
SMILES C1=CC=C2C(=C1)C(=O)NS2(=O)=O.O.O.[Na]

Specifications

Alkalinity Pass Test (FCC)
Acidity Pass Test (USP)
Color White
Packaging Poly Bottle
Quantity 2.5 kg
Residual Solvents Meets Requirements (USP)
Formula Weight 205.17
Percent Purity 98.0 to 101.0% (USP)
Grade FCC/USP
Physical Form Solid
Chemical Name or Material Saccharin Sodium
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.