Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saccharomyces Cerevisiae Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10066314 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10066314 0.5 mL
NB017765 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10066314 Supplier Novus Biologicals Supplier No. NB10066314
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Saccharomyces Cerevisiae Polyclonal specifically detects Saccharomyces Cerevisiae in Yeast samples. It is validated for Western Blot.

Specifications

Antigen Saccharomyces Cerevisiae
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation Phosphate Buffered Saline with 0.09% Sodium Azide
Gene Alias Bakers yeast, Baker's yeast, Bakers' yeast, Brewers yeast, Brewer's yeast, Brewers' yeast, S cerevisiae, S. cerevisiae, S.cerevisiae
Host Species Rabbit
Immunogen Whole cells of S. cerevisiae.
Purification Method IgG purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-66314 recognizes Saccharomyces cerevisiae, a rapidly growing budding yeast commonly used in brewing and baking. S. cerevisiae has been studied intensively as a eukaryotic cell model as it is easy to culture but maintains the internal complexity of plant and animal cells.
Target Species Yeast
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.