Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Saccharomyces Cerevisiae Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $560.50
Specifications
Antigen | Saccharomyces Cerevisiae |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017765
![]() |
Novus Biologicals
NB100663140.1ML |
0.1 mL |
Each for $206.50
|
|
|||||
NB10066314
![]() |
Novus Biologicals
NB10066314 |
0.5 mL |
Each for $560.50
|
|
|||||
Description
Saccharomyces Cerevisiae Polyclonal specifically detects Saccharomyces Cerevisiae in Yeast samples. It is validated for Western Blot.Specifications
Saccharomyces Cerevisiae | |
Western Blot 1:100-1:2000 | |
Polyclonal | |
Purified | |
RUO | |
Phosphate Buffered Saline with 0.09% Sodium Azide | |
Whole cells of S. cerevisiae. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Yeast | |
Bakers yeast, Baker's yeast, Bakers' yeast, Brewers yeast, Brewer's yeast, Brewers' yeast, S cerevisiae, S. cerevisiae, S.cerevisiae | |
IgG | |
IgG purified | |
NB100-66314 recognizes Saccharomyces cerevisiae, a rapidly growing budding yeast commonly used in brewing and baking. S. cerevisiae has been studied intensively as a eukaryotic cell model as it is easy to culture but maintains the internal complexity of plant and animal cells. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title