Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SACM1L Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB330707 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB330707 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB330707 Supplier Novus Biologicals Supplier No. NBP297655APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SACM1L Polyclonal antibody specifically detects SACM1L in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen SACM1L
Applications Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias DKFZp686A0231, EC 3.1.3, EC 3.1.3.36, EC 3.1.3.77, KIAA0851suppressor of actin 1, phosphatidylinositide phosphatase SAC1, SAC1 suppressor of actin mutations 1-like (yeast), SAC1EC 3.1.3.-, Suppressor of actin mutations 1-like protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SACM1L fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 22908
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.