Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SACM1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106374
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106374 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106374 Supplier Invitrogen™ Supplier No. PA5106374
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SACM1L.

Phosphoinositide phosphatase that hydrolyzes PtdIns(3)P and PtdIns(4)P. Has low activity towards PtdIns(3,5)P2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SACM1L
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Sacm1l
Gene Accession No. Q9EP69, Q9ES21, Q9NTJ5
Gene Alias Kiaa0851; mKIAA0851; phosphatidylinositide phosphatase SAC1; phosphatidylinositol-3-phosphatase SAC1; phosphatidylinositide phosphatase SAC1; phosphatidylinositol-4-phosphate phosphatase; polyphosphatidylinositol phosphatase; SAC1; SAC1 (suppressor of actin mutations 1, homolog)-like (S. cerevisiae); SAC1 like phosphatidylinositide phosphatase; SAC1 suppressor of actin mutations 1 like; SAC1 suppressor of actin mutations 1-like (yeast); Sac1p; SACM1L; suppressor of actin 1; suppressor of actin mutations 1-like protein
Gene Symbols Sacm1l
Host Species Rabbit
Immunogen A synthesized peptide derived from human SACM1L(Accession Q9NTJ5), corresponding to amino acid residues R267-L317.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116482, 22908, 83493
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.