Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAE2 Antibody (OTI4A3), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP273973V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273973V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273973V Supplier Novus Biologicals Supplier No. NBP273973V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SAE2 Monoclonal specifically detects SAE2 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SAE2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4A3
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Anthracycline-associated resistance ARX, ARX, EC 6.3.2, EC 6.3.2.-, FLJ13058, HRIHFB2115, SAE2SUMO-1 activating enzyme subunit 2, SUMO1 activating enzyme subunit 2, SUMO-activating enzyme subunit 2, UBA2, ubiquitin-activating enzyme E1 homolog, Ubiquitin-like 1-activating enzyme E1B, ubiquitin-like modifier activating enzyme 2, UBLE1B
Gene Symbols UBA2
Host Species Mouse
Immunogen Full length recombinant protein of human SAE2 (NP_005490) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10054.0
Target Species Human, Mouse, Rat, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.