Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab™ SafeTech Vapor Monitor
SDP

Catalog No. STLVMBISOP
Click to view available options
Components Measured:
Ethylene Oxide
Formaldehyde
Isopropanol
Xylene

Catalog No. STLVMBISOP

Supplier: StatLab™ VMBISOP

Vapor monitoring is important to ensure the safety of employees and comply with OSHA guidelines.

  • StatLab™ makes it easy to monitor vapor levels in your laboratory
  • Simply send the badge in and access your results via our website
  • Includes prepaid analysis for 8-hour or 15-minute exposure readings
  • Complies with OSHA standards for vapor exposure in the workplace

Specifications

Components Measured Isopropanol
For Use With (Equipment) Laboratory
Type Single Gas Detector
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.