Learn More
StatLab™ SafeTech Vapor Monitor

Description
- StatLab™ makes it easy to monitor vapor levels in your laboratory
- Simply send the badge in and access your results via our website
- Includes prepaid analysis for 8-hour or 15-minute exposure readings
- Complies with OSHA standards for vapor exposure in the workplace
Specifications
Specifications
| Components Measured | Xylene |
| For Use With (Equipment) | Laboratory |
| Type | Single Gas Detector |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.