Learn More
Safety Red-Spirit™ Thermometers
SCI_ED
Catalog No. S41575D
Click to view available options
:
-10 to + 200 deg. C
-10 to +200 deg. C/ Sub
This item is not returnable. View return policy
This item is not returnable. View return policy
Solid-stem glass thermometers with dark scale and white background are easy for students to read and can withstand heavy usage.
  • Unique, anti-roll device reduces accidental breakage
  • Individually wrapped in safety seal packs
  • Models with PTFE coating encapsulate the glass and liquid in the event of breakage
  • Premium-grade thermometers have superior quality black markings
  • Student-grade thermometers, most commonly used in school laboratories, have dark brown markings
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.