Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SAG
GREENER_CHOICE

Catalog No. p-200038197
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent Smoothened receptor agonist; activates the Hedgehog signaling pathway

SAG is a potent Smoothened (Smo) receptor agonist (Kd = 59 nM); antagonizes Cyclopamine (Cat. No. 1623) action at the Smo receptor. SAG potently activates the Hedgehog signaling pathway in Shh-light 2 cells (EC50 ∼ 3 nM) and nduces pathway activation independently of Ptch proteins. SAG is a putative inhibitor of a cellular component required for Hedgehog signaling and also enhances neuronal differentiation of iPSCs into dopaminergic neurons.

SAG dihydrochloride (Cat. No. 6390) also available.

Specifications

Quantity 1 mg
Product Type SAG
Purity >99%
Molecular Weight (g/mol) 490.06
Inhibitors SAG

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.