Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAH3 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB292069 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292069 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292069 Supplier Novus Biologicals Supplier No. NB100400APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SAH3 Polyclonal antibody specifically detects SAH3 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen SAH3
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias adenosylhomocysteinase-like 2, AdoHcyase 3, ADOHCYASE3, EC 3.3.1.1, FLJ21719, KIAA0828S-adenosylhomocysteine hydrolase-like 2, putative adenosylhomocysteinase 3, S-adenosylhomocysteine hydrolase-like protein 2, S-adenosyl-L-homocysteine hydrolase 3
Host Species Rabbit
Immunogen Maps to a region between residues 1 and 50 of human adenosylhomocysteinase 3 (KIAA0828) using the numbering given in entry NP_056143.1 (GeneID 23382).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23382
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.