Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SAHA
GREENER_CHOICE

Catalog No. p-4817110
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Class I and II HDAC inhibitor

SAHA inhibits Class I and II histone deacetylases (HDACs); induces accumulation of acetylated histones H2A, H2B, H3 and H4 in transformed cultured cells. Suppresses cell growth in a range of cancer cell lines; induces apoptosis in cutaneous T cell lymphoma cells in vitro. Activates autophagy. SAHA increases efficiency of transcription factor-induced reprogramming of mouse embryonic fibroblasts (MEF) to induced pluripotent stem cells (iPSC). Also enhances adeno-associated virus transduction of HeLa cells.

Chemical Identifiers

CAS 149647-78-9
Molecular Formula C14H20N2O3

Specifications

Quantity 10 mg
Formula Weight 264.32
Purity >98%
Molecular Weight (g/mol) 264.325
Chemical Name or Material SAHA
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.