Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SAK 3

Click to view available options
Quantity:
5 mg
25 mg
Description
SAK 3 is a potent CaV3.1 and CaV3.3 activator. Enhances CaV3.1 and CaV3.3, but not CaV3.2 activity at concentrations of 0.01 - 10 M in neuro2A cells in vitro. Elevates ACh levels in hippocampus. Rescues impaired memory-related behaviors, promotes adult neurogenesis and displays antidepressant-like effects in OBX mice. Orally bioavailable.
Specifications
Specifications
Chemical Name or Material | Ethyl 8′-methyl-2′,4-dioxo-2-(piperidin-1-yl)-2′H-spiro[cyclopent[2]ene-1,3′-imidazo[1,2-a]pyridine]-3-carboxylate |
CAS | 1256269-87-0 |
Quantity | 5 mg |
Target | Cav3.x Channel Activators |
Molecular Formula | C20H23N3O4 |
Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction