Learn More
Salicylaldehyde, 99%
Description
Precursor to a variety of chelating agents.Salicylaldehyde is used as a precursor to prepare catechol, benzofuran, salicylaldehydimine and 3-carbethoxycoumarin. It is used for colorimetric assay of isoleucine as well as utilized for the determination of primary amine by fluorescence method. It is a key starting material to a various chelating agents. It is used as flavor and fragrance components. Its derivative salicylaldehyde thiosemicarbazone is used for the simultaneous determination of molybdenum and iron by spectrophotometric method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Precursor to a variety of chelating agents.Salicylaldehyde is used as a precursor to prepare catechol, benzofuran, salicylaldehydimine and 3-carbethoxycoumarin. It is used for colorimetric assay of isoleucine as well as utilized for the determination of primary amine by fluorescence method. It is a key starting material to a various chelating agents. It is used as flavor and fragrance components. Its derivative salicylaldehyde thiosemicarbazone is used for the simultaneous determination of molybdenum and iron by spectrophotometric method.
Solubility
Miscible with alcohol, ether, acetone, benzene and most organic solvents. Slightly miscible with water.
Notes
Incompatible with acids, strong reducing agents, strong oxidizing agents, strong bases and halogens.
Specifications
Specifications
| Melting Point | °C to 7°C |
| Density | 1.168 |
| Boiling Point | 197°C |
| Flash Point | 76°C (168°F) |
| Assay Percent Range | 99% |
| Refractive Index | 1.573 |
| Quantity | 100 g |
| Beilstein | 471388 |
| Sensitivity | Air and light sensitive |
| Merck Index | 14,8326 |
| Show More |
Safety and Handling
| Hazard Category | H227-H302-H311-H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335-H227 |
| Precautionary Statement | P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P309+P311-P312-P314-P330-P332+P313-P361-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: III |
| EINECSNumber | 201-961-0 |
| RTECSNumber | VN5250000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.