Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Salivary Amylase Alpha Antibody (1G4D8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261869
Description
Salivary Amylase Alpha Monoclonal specifically detects Salivary Amylase Alpha in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
Salivary Amylase Alpha | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
1,4-alpha-D-glucan glucanohydrolase 1, alpha-amylase 1, AMY1, AMY1B, AMY1C, amylase, alpha 1A (salivary), amylase, alpha 1A; salivary, amylase, salivary, alpha-1A, EC 3.2.1.1, glycogenase, Salivary alpha-amylase, salivary amylase alpha 1A | |
Mouse | |
57.8 kDa | |
0.1 mL | |
276 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, ELISA | |
1G4D8 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
AMY1A | |
Purified recombinant fragment of human AMY1A (AA: 172-284) expressed in E. Coli. | |
Protein G purified | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction