Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Salivary Amylase Alpha Antibody (1G4D8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$562.50
Specifications
Antigen | Salivary Amylase Alpha |
---|---|
Clone | 1G4D8 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
Applications | Western Blot, Flow Cytometry, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61869
![]() |
Novus Biologicals
NBP261869 |
0.1 mL |
Each of 1 for $562.50
|
|
|||||
Description
Salivary Amylase Alpha Monoclonal specifically detects Salivary Amylase Alpha in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
Salivary Amylase Alpha | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Mouse | |
276 | |
Purified recombinant fragment of human AMY1A (AA: 172-284) expressed in E. Coli. | |
Primary | |
57.8 kDa |
1G4D8 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
1,4-alpha-D-glucan glucanohydrolase 1, alpha-amylase 1, AMY1, AMY1B, AMY1C, amylase, alpha 1A (salivary), amylase, alpha 1A; salivary, amylase, salivary, alpha-1A, EC 3.2.1.1, glycogenase, Salivary alpha-amylase, salivary amylase alpha 1A | |
AMY1A | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title