Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SALL1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP213275 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP213275 0.1 mL
NB438617 25ul
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP213275 Supplier Novus Biologicals Supplier No. NBP213275
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SALL1 Polyclonal specifically detects SALL1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SALL1
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias HSAL1, sal (Drosophila)-like 1, Sal-1, SAL1, sal-like 1 (Drosophila), sal-like protein 1, Spalt-like transcription factor 1, Zinc finger protein 794, Zinc finger protein SALL1, Zinc finger protein Spalt-1, ZNF794TBS
Gene Symbols SALL1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the amino acids: TLPSLIPFIKTEEPAPIPISHSATSPPGSVKSDSGGPESATRNLGGLPEEAEGSTLPPSGGKSEESGMVTNSVPT
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6299
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.