Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SALL1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP152141 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152141 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152141 Supplier Novus Biologicals Supplier No. NBP152141
Add to Cart
Add to Cart

Goat Polyclonal Antibody

SALL1 Polyclonal specifically detects SALL1 in Human, Mouse samples. It is validated for Peptide ELISA.

Specifications

Antigen SALL1
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_002959.2
Gene Alias HSAL1, sal (Drosophila)-like 1, Sal-1, SAL1, sal-like 1 (Drosophila), sal-like protein 1, Spalt-like transcription factor 1, Zinc finger protein 794, Zinc finger protein SALL1, Zinc finger protein Spalt-1, ZNF794TBS
Gene Symbols SALL1
Host Species Goat
Immunogen Peptide with sequence EKGQPSRPTKSKDAH-C corresponding to N-Terminus according to NP_002959.2, NP_001121364.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6299
Test Specificity This antibody is expected to recognize both reported isoforms (NP_002959.2; NP_001121364.1).
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.