Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SALM4/LRFN3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB396510 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396510 0.025 mg
NBP241098 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB396510 Supplier Novus Biologicals Supplier No. NBP2410980.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SALM4/LRFN3 Polyclonal specifically detects SALM4/LRFN3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SALM4/LRFN3
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% Sodium Azide
Gene Alias fibronectin type III, immunoglobulin and leucine rich repeat domains 1, FIGLER1, leucine rich repeat and fibronectin type III domain containing 3, leucine-rich repeat and fibronectin type-III domain-containing protein 3, MGC2656, SALM4, synaptic adhesion-like molecule 4, UNQ5865/PRO34192
Gene Symbols LRFN3
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the center of human LRFN3. The immunogen is located within amino acids 380 - 430 of LRFN3. Amino Acid Squence: DPPRDGDPDALTPPS
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79414
Test Specificity This antibody is predicted to not cross-react with other members of the LRFN family.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.