Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salmonella Antibody (100/358), Biotin, Novus Biologicals™
SDP

Catalog No. NB211674 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB211674 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB211674 Supplier Novus Biologicals Supplier No. NB10064563B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Salmonella Monoclonal antibody specifically detects Salmonella in Bacteria samples. It is validated for ELISA, Immunofluorescence

Specifications

Antigen Salmonella
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 100/358
Conjugate Biotin
Formulation PBS
Gene Alias Salmonellae
Host Species Mouse
Immunogen Native S. enteritidis (NCTC 5188 9.12:g,m).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.