Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salmonella Enteritidis Antibody (10D9H (E91)), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N1066471A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1066471A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1066471A70 Supplier Novus Biologicals Supplier No. NB10066471AF750

Mouse Monoclonal Antibody

Salmonella Enteritidis Monoclonal specifically detects Salmonella Enteritidis in Bacteria samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Salmonella Enteritidis
Applications Western Blot, ELISA
Classification Monoclonal
Clone 10D9H (E91)
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Host Species Mouse
Immunogen Heat inactivated cells from Salmonella (LPS groups A, B & D).
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity This antibody recognizes a common antigenic determinant that is found in the LPS of Salmonella enteriditis (Group D), Salmonella typhimurium (Group B) and Salmonella paratyphi (Group A) . This antibody is thought to recognize the ′0-12′ antigen [2-D-mannose (1-4) I- ramnose (1-3)-D galactose] of salmonella LPS. A similar sequence is found in Group E, which may account for the weak cross reactivity observed with bacteria of this group.
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.