Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Salmonella Enteritidis Antibody (10D9H (E91)), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100664710.025MG
Description
Salmonella Enteritidis Monoclonal specifically detects Salmonella Enteritidis in Bacteria samples. It is validated for Western Blot, ELISA.Specifications
Salmonella Enteritidis | |
Monoclonal | |
Unconjugated | |
PBS with 0.09% Sodium Azide | |
Heat inactivated cells from Salmonella (LPS groups A, B & D). | |
0.025 mg | |
Primary | |
Bacteria | |
IgG1 |
ELISA, Western Blot | |
10D9H (E91) | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Mouse | |
Affinity Purified | |
RUO | |
This antibody recognizes a common antigenic determinant that is found in the LPS of Salmonella enteriditis (Group D), Salmonella typhimurium (Group B) and Salmonella paratyphi (Group A) . This antibody is thought to recognize the '0-12' antigen [2-D-mannose (1-4) I- ramnose (1-3)-D galactose] of salmonella LPS. A similar sequence is found in Group E, which may account for the weak cross reactivity observed with bacteria of this group. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction