Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Salmonella Enteritidis Antibody (10D9H (E91)), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
Antigen | Salmonella Enteritidis |
---|---|
Clone | 10D9H (E91) |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | ELISA, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017775
![]() |
Novus Biologicals
NB100664710.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10066471
![]() |
Novus Biologicals
NB10066471 |
0.1 mg | N/A | N/A | N/A | ||||
Description
Salmonella Enteritidis Monoclonal specifically detects Salmonella Enteritidis in Bacteria samples. It is validated for Western Blot, ELISA.Specifications
Salmonella Enteritidis | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Mouse | |
Bacteria | |
Heat inactivated cells from Salmonella (LPS groups A, B & D). | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
10D9H (E91) | |
ELISA, Western Blot | |
Unconjugated | |
RUO | |
PBS with 0.09% Sodium Azide | |
IgG1 | |
Affinity Purified | |
This antibody recognizes a common antigenic determinant that is found in the LPS of Salmonella enteriditis (Group D), Salmonella typhimurium (Group B) and Salmonella paratyphi (Group A) . This antibody is thought to recognize the '0-12' antigen [2-D-mannose (1-4) I- ramnose (1-3)-D galactose] of salmonella LPS. A similar sequence is found in Group E, which may account for the weak cross reactivity observed with bacteria of this group. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title