Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Salmonella Antibody (100/358) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$181.90 - $487.50
Specifications
| Antigen | Salmonella |
|---|---|
| Clone | 100/358 |
| Concentration | 1.0 mg/mL |
| Dilution | ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
| Applications | Immunocytochemistry, Immunofluorescence, ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017611
![]() |
Novus Biologicals
NB100645630.025MG |
0.025 mg |
Each for $181.90
|
|
|||||
NB10064563
![]() |
Novus Biologicals
NB10064563 |
0.1 mg |
Each for $487.50
|
|
|||||
Description
Salmonella Monoclonal specifically detects Salmonella in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Salmonella | |
| 1.0 mg/mL | |
| Immunocytochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Mouse | |
| Bacteria | |
| IgG2a | |
| Protein A purified | |
| Antibody recognizes an antigen common to the various Salmonella strains. |
| 100/358 | |
| ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Native S. enteritidis (NCTC 5188 9.12:g,m). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title