Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salmonella Paratyphi A Antibody (10B10G), Novus Biologicals™
SDP

Catalog No. NB287550 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287550 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287550 Supplier Novus Biologicals Supplier No. NB2004070.2MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Salmonella Paratyphi A Monoclonal antibody specifically detects Salmonella Paratyphi A in Bacteria samples. It is validated for Western Blot

Specifications

Antigen Salmonella Paratyphi A
Applications Western Blot
Classification Monoclonal
Clone 10B10G
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS (pH 7.4)
Gene Alias Salmonellae
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with heat inactivated bacterial cells of Salmonella paratyphi A.
Purification Method Protein G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Cell Biology, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.