Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Salmonella typhi Mouse anti-All, Unconjugated, Clone: Clone 4, MilliporeSigma™
Mouse Monoclonal Antibody
$955.79 - $9553.82
Specifications
Antigen | Salmonella typhi |
---|---|
Clone | Clone 4 |
Applications | ELISA, Immunodiffusion, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABX87541MI
|
MilliporeSigma
MABX87541KC |
1 mg |
Each for $955.79
|
|
|||||
MABX87410MI
|
MilliporeSigma
MABX875410KC |
10 mg |
Each for $9,553.82
|
|
|||||
Description
Salmonella typhi Monoclonal antibody specifically detects Salmonella typhi in All samples. It is validated for Enzyme Immunoassay (ELISA), Immunodiffusion, Western Blotting.Specifications
Salmonella typhi | |
ELISA, Immunodiffusion, Western Blot | |
Unconjugated | |
Infectious Diseases | |
Anti-S. typhi | |
IgG2b | |
Affinity Purified | |
Specific for S. Typhi and S. Paratyphi |
Clone 4 | |
Monoclonal | |
Mouse | |
All species | |
Negative S. typhi Antigen | |
Primary | |
2 to 8°C. For long-term storage, freeze at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title