Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAMD12 Antibody (OTI2G1), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N273974J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273974J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273974J646 Supplier Novus Biologicals Supplier No. NBP273974JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SAMD12 Monoclonal specifically detects SAMD12 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SAMD12
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2G1
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ39458, MGC148139, MGC148140, SAM domain-containing protein 12, sterile alpha motif domain containing 12, sterile alpha motif domain-containing protein 12
Gene Symbols SAMD12
Host Species Mouse
Immunogen Full length recombinant protein of human SAMD12 (NP_997389) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 401474.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.