Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAMD8 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen SAMD8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP1601320
SDP
View Documents
Novus Biologicals
NBP16011320UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP160113
SDP
View Documents
Novus Biologicals
NBP160113
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

SAMD8 Polyclonal specifically detects SAMD8 in Human samples. It is validated for Western Blot.
Specifications

Specifications

SAMD8
Polyclonal
Rabbit
Lipid and Metabolism
FLJ25082, SAM domain-containing protein 8, SMSr, sphingomyelin synthase related, sphingomyelin synthase-related protein 1, sterile alpha motif domain containing 8, Sterile alpha motif domain-containing protein 8
SAMD8
IgG
Western Blot
Unconjugated
RUO
Q5JSC8
142891
Synthetic peptides corresponding to SAMD8(sterile alpha motif domain containing 8) The peptide sequence was selected from the middle region of SAMD8 (NP_653261). Peptide sequence MQTYPPLPDIFLDSVPRIPWAFAMTEVCGMILCYIWLLVLLLHKHRSILL.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.