Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SAMHD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP227097SS
Description
SAMHD1 Polyclonal specifically detects SAMHD1 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SAMHD1 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| SAMHD1 | |
| A portion of amino acids 1-50 from human SAMHD1 was used as the immunogen for this antibody. | |
| 0.025 mg | |
| Stem Cell Markers | |
| 25939 | |
| Human, Primate | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 2-4ug/ml, Immunohistochemistry-Paraffin 5ug/ml | |
| DCIPMg11, Dendritic cell-derived IFNG-induced protein, EC 3.1.4.-, HDDC1, Monocyte protein 5, MOP5, MOP-5AGS5, SAM domain and HD domain 1, SAM domain and HD domain-containing protein 1, SBBI88 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human, orangutan, chimpanzee, and white-cheeked gibbon, 93% in rhesus monkey, dog, horse, and mole rat, 86% in guinea pig, new world monkey, Chinese hamster, and common rabbit, 79% in panda, macaque, cat, and rat, 71% in african bush elephant, and 57% homologous in mouse. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction