Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAMHD1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$149.33 - $394.26

Specifications

Antigen SAMHD1
Concentration 0.5 mg/mL
Dilution Western Blot 2-4ug/ml, Immunohistochemistry-Paraffin 5ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227097SS
SDP
View Documents
Novus Biologicals
NBP227097SS
0.025 mg
Each for $149.33
Only null left
Add to Cart
 
NBP227097
SDP
View Documents
Novus Biologicals
NBP227097
0.1 mg
Each for $394.26
Only null left
Add to Cart
 
Description

Description

SAMHD1 Polyclonal specifically detects SAMHD1 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

SAMHD1
Western Blot 2-4ug/ml, Immunohistochemistry-Paraffin 5ug/ml
Polyclonal
Purified
RUO
Human, Primate
DCIPMg11, Dendritic cell-derived IFNG-induced protein, EC 3.1.4.-, HDDC1, Monocyte protein 5, MOP5, MOP-5AGS5, SAM domain and HD domain 1, SAM domain and HD domain-containing protein 1, SBBI88
SAMHD1
IgG
Protein A purified
The amino acid sequence used as immunogen for this antibody is 100% homologous in human, orangutan, chimpanzee, and white-cheeked gibbon, 93% in rhesus monkey, dog, horse, and mole rat, 86% in guinea pig, new world monkey, Chinese hamster, and common rabbit, 79% in panda, macaque, cat, and rat, 71% in african bush elephant, and 57% homologous in mouse.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Stem Cell Markers
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
25939
A portion of amino acids 1-50 from human SAMHD1 was used as the immunogen for this antibody.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.