Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAMM50 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB134979 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB134979 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB134979 Supplier Novus Biologicals Supplier No. NBP306283G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SAMM50 Polyclonal specifically detects SAMM50 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SAMM50
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CGI-51, FLJ42905, FLJ99036, OMP85, SAM50, sorting and assembly machinery 50kDa, sorting and assembly machinery component 50 homolog, sorting and assembly machinery component 50 homolog (S. cerevisiae), TOB55, Transformation-related gene 3 protein, transformation-related protein 3, TRG-3FLJ35825, YNL026W
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SAMM50 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 25813
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.