Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ Sampling Hoses and Tools for Dräger™ accuro™ Bellows Pumps

Catalog No. p-177133 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Length (Metric):
10 m
15 m
3 m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
17-988-413 3 m
17-988-414 10 m
19-453-030 15 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-988-413 Supplier Dräger™ Supplier No. 6400077
Only null left
Add to Cart
Add to Cart

Specifications

For Use With (Equipment) Accuro Bellows Pump
Type Extension Hose
Includes Adapter
Length (English) 10 ft.
Length (Metric) 3 m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.