Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ITL BioMedical SampLok™ Adapter Cap 2
Click to view available options
Quantity:
480/Cs., 40/Box, 12 Boxes/Cs.
Description
Compliance
CE marked
Specifications
Specifications
Pre-sterilized | No |
Product Type | Blood Collection Adapter |
Autoclavable | Autoclavable |
Description | Without luer |
Material | Polypropylene |
Color | Clear |
For Use With (Application) | Blood Collection |
Quantity | 480/Cs., 40/Box, 12 Boxes/Cs. |
Sterility | Non-sterile |
For Use With (Equipment) | Blood Collection Needles, Blood Culture Bottle, Blood Collection Tubes (A variety of blood collection and winged needle sets can be attached) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction